missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) ammonium nitrate tetrahydrate, Reagent Grade, Thermo Scientific Chemicals

£54.20 - £151.00

Chemical Identifiers

CAS 13083-04-0
Molecular Formula CeH16N7O19
Molecular Weight (g/mol) 558.27
MDL Number MFCD00213483
InChI Key AIVZRMAXALRCHB-UHFFFAOYSA-N
Synonym ce.3no3.2hno3.4h2o.2nh3, ammonium cerium iii nitrate tetrahydrate, cerous ammonium nitrate, hydrated, reagent, cerium iii ammonium nitrate tetrahydrate, reagent grade, cerium 3+ bis nitric acid diamine tetrahydrate trinitrate
PubChem CID 91886599
IUPAC Name azane;cerium(3+);nitric acid;trinitrate;tetrahydrate
SMILES N.N.O.O.O.O.[Ce+3].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11304467
View Documents Promotion Details
Thermo Scientific Alfa Aesar
038367.22
100 g
£54.20
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11314467
View Documents Promotion Details
Thermo Scientific Alfa Aesar
038367.36
500 g
£151.00
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A useful oxidant for organic synthesis. This chemical is also useful for the synthesis of quinoxaline derivatives which are useful as DNA cleaving agents, dyes, and organic semiconductors. Used to release organic ligands from metal carbonyls. Whereby the metal is oxidized, Carbon monoxide is produced, and the organic ligand is released.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13083-04-0
558.27
AIVZRMAXALRCHB-UHFFFAOYSA-N
91886599
N.N.O.O.O.O.[Ce+3].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
CeH16N7O19
MFCD00213483
ce.3no3.2hno3.4h2o.2nh3, ammonium cerium iii nitrate tetrahydrate, cerous ammonium nitrate, hydrated, reagent, cerium iii ammonium nitrate tetrahydrate, reagent grade, cerium 3+ bis nitric acid diamine tetrahydrate trinitrate
azane;cerium(3+);nitric acid;trinitrate;tetrahydrate

Specifications

13083-04-0
Crystalline
MFCD00213483
Soluble in water.
N.N.O.O.O.O.[Ce+3].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
558.27
558.28 (486.22 Anhydrous)
White crystalline
White
CeH16N7O19
ce.3no3.2hno3.4h2o.2nh3, ammonium cerium iii nitrate tetrahydrate, cerous ammonium nitrate, hydrated, reagent, cerium iii ammonium nitrate tetrahydrate, reagent grade, cerium 3+ bis nitric acid diamine tetrahydrate trinitrate
AIVZRMAXALRCHB-UHFFFAOYSA-N
azane;cerium(3+);nitric acid;trinitrate;tetrahydrate
91886599
Reagent
Cerium(III) ammonium nitrate tetrahydrate, For reagent grade
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c

H272-H315-H319

missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: III

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only