missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) acetate sesquihydrate, 99.9% (REO)

Product Code. 11307118
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307118 100 g 100g
11317118 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307118 Supplier Thermo Scientific Alfa Aesar Supplier No. 015294.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

With bromide ion, catalyzes the liquid-phase auto-oxidation of cresols | CAS: 537-00-8 | C6H9CeO6 | 317.248 g/mol

With bromide ion, catalyzes the liquid-phase auto-oxidation of cresols.Cerium(III) acetate sesquihydrate, acts as a source of cerium involved in the preparation of cerium oxide. It finds application in metallurgy, glass and glass polishing, ceramics, catalysts, and in phosphors. It is utilized in steel production to remove the free sulfur and oxygen. Further, it is an efficient glass polishing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 537-00-8
Molecular Formula C6H9CeO6
Molecular Weight (g/mol) 317.248
MDL Number MFCD00051060
InChI Key VGBWDOLBWVJTRZ-UHFFFAOYSA-K
Synonym cerium acetate, cerous acetate, cerium 3+ acetate, cerium iii acetate, cerium triacetate, cerium 3+ triacetate, unii-yyb5291r2h, acetic acid, cerium 3+ salt, cerium iii acetate sesquihydrate
PubChem CID 10826
IUPAC Name cerium(3+);triacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Ce+3]

Specifications

Melting Point 308°C (decomposition)
Color White
Physical Form Powder
Quantity 100 g
Solubility Information Soluble in water.
Formula Weight 344.28 (317.26 Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Cerium(III) acetate sesquihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.