missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) acetate hydrate, 99.995% (metals basis)

Product Code. 11430730
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430730 10 g 10g
11440730 50 g 50g
11450730 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430730 Supplier Thermo Scientific Alfa Aesar Supplier No. 044433.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 206996-60-3 | C6H9CeO6 | 317.25 g/mol

Cerium(III) acetate hydrate is used in thin fim buffer layers suitable for high κ dielectrics by sol-gel route. And it is also used as a catalyst for auto-oxidation of cresols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206996-60-3
Molecular Formula C6H9CeO6
Molecular Weight (g/mol) 317.25
MDL Number MFCD00051060
InChI Key VGBWDOLBWVJTRZ-UHFFFAOYSA-K
IUPAC Name cerium(3+) triacetate
SMILES [Ce+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Color White
Physical Form Crystalline
Quantity 10 g
Linear Formula Ce(C2H3O2)3·xH2O
Solubility Information Soluble in water( 3.5g/L).
Formula Weight 317.26 (Anhydrous)
Percent Purity 99.995%
Assay (metals basis)
Chemical Name or Material Cerium (III) acetate hydrate
compliance-icons compliance-icons
Product Identifier
  • Cerium(III) acetate hydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Cerium(III) acetatehydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.