missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) 2,4-pentanedionate hydrate, Thermo Scientific Chemicals

Catalog Number 11347997
112.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
206996-61-4
C15H21CeO6
437.44
MFCD00150164
LCRQZRNVLFDPAZ-UHFFFAOYSA-N
cerium iii acetylacetonate hydrate, cerium iii 2,4-pentanedionate, cerium iii 2,4-pentanedionate hydrate, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy cerio oxy pent-3-en-2-one hydrate
16212434
[Ce+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
206996-61-4
C15H21CeO6
437.44
MFCD00150164
LCRQZRNVLFDPAZ-UHFFFAOYSA-N
cerium iii acetylacetonate hydrate, cerium iii 2,4-pentanedionate, cerium iii 2,4-pentanedionate hydrate, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy cerio oxy pent-3-en-2-one hydrate
16212434
[Ce+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Cerium(III) 2,4-pentanedionate hydrate used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 131°C to 132°C
Color Red to Yellow
Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water.
IUPAC Name cerium;(Z)-4-hydroxypent-3-en-2-one;hydrate
Formula Weight 437.44 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Cerium(III) 2,4-pentanedionate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'