missing translation for 'onlineSavingsMsg'
Learn More

Cerium Chloride, TRC

Product Code. 30351117
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351117 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351117 Supplier Toronto Research Chemicals Supplier No. TRCC27855025G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cerium Chloride
CAS 7790-86-5
Molecular Formula CeCl3
Quantity 25 g
Synonym Cerium Trichloride, Cerium(III) Chloride, Cerous Chloride, Cerous(III) Chloride, NSC 84267
InChI Formula InChI=1S/Ce.3ClH/h;3*1H/q+3;;;/p-3
SMILES Cl[Ce](Cl)Cl
IUPAC Name trichlorocerium
Molecular Weight (g/mol) 246.48
Formula Weight 244.812
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.