missing translation for 'onlineSavingsMsg'
Learn More

Ceritinib, Thermo Scientific™

Product Code. 17770649 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17770649 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17770649 Supplier Thermo Scientific Acros Supplier No. 469322500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material Ceritinib
CAS 1032900-25-6
Quantity 250 mg
Melting Point 172°C to 177°C
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C28 H36 Cl N5 O3 S
Physical Form Powder
Color White
Purity ≥97.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.