missing translation for 'onlineSavingsMsg'
Learn More

Ceric Ammonium Sulfate Dihydrate, TRC

Product Code. 30350224
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30350224 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350224 Supplier Toronto Research Chemicals Supplier No. TRCC2625705G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonium cerium sulfate
CAS 10378-47-9
Molecular Formula Ce . 4 H O4 S . 2 H2 O . 4 H3 N
Quantity 5 g
Synonym Diammonium cerium(IV) sulphate
InChI Formula InChI=1S/Ce.4H3N.4H2O4S.2H2O/c;;;;;4*1-5(2,3)4;;/h;4*1H3;4*(H2,1,2,3,4);2*1H2/q+4;;;;;;;;;;/p-4
SMILES N.N.N.N.O.O.[Ce+4].OS(=O)(=O)[O-].OS(=O)(=O)[O-].OS(=O)(=O)[O-].OS(=O)(=O)[O-]
IUPAC Name azane;cerium(4+);hydrogen sulfate;dihydrate
Molecular Weight (g/mol) 632.55
Formula Weight 631.871
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.