missing translation for 'onlineSavingsMsg'
Learn More

Ceric Ammonium Nitrate, ACS Reagent Grade, Ricca Chemical

Product Code. 31750140
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31750140 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31750140 Supplier Ricca Chemical Company Supplier No. RDCC0300500B1
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 16774-21-3
CAS Min % 98.0
CAS Max % 100.0
Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Molecular Weight (g/mol) 548.26 g/mol
Percent Purity ≥ 98.5%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Ceric Ammonium Nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.