missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cerebral Protein 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558412
This item is not returnable.
View return policy
Description
Immunogen sequence: AQRLFQEDPE AFLLYHRGFQ SQVKKWPLQP VDRIARDLRQ RPASLVVADF GCGDCRLASS IRNSVHCFDL ASLDPRVTVC DMAQVPLEDE SVDVAVFC Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
Essential component of the eNoSC (energy-dependent nucleolar silencing) complex, a complex that mediates silencing of rDNA in response to intracellular energy status and acts by recruiting histone-modifying enzymes. The eNoSC complex is able to sense the energy status of cell: upon glucose starvation, elevation of NAD(+)/NADP(+) ratio activates SIRT1, leading to histone H3 deacetylation followed by dimethylation of H3 at 'Lys-9' (H3K9me2) by SUV39H1 and the formation of silent chromatin in the rDNA locus. In the complex, RRP8 binds to H3K9me2 and probably acts as a methyltransferase. Its substrates are however unknown.
Specifications
| Cerebral Protein 1 | |
| Polyclonal | |
| Unconjugated | |
| Rrp8 | |
| 1500003O22Rik; 2900001K19Rik; AW538116; Cerebral protein 1; cerebral protein 1 homolog; HUCE1; hucep1; hucep-1; KIAA0409; NML; Nucleomethylin; RGD1308302; ribosomal RNA processing 8, methyltransferase, homolog (yeast); ribosomal RNA-processing protein 8; Rrp8; RRP-8; RRP8 methyltransferase homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23378 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43159 | |
| Rrp8 | |
| Recombinant protein corresponding to Human Cerebral Protein 1. Recombinant protein control fragment (Product #RP-97327). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction