missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cerebellin-4 Polyclonal antibody specifically detects Cerebellin-4 in Human samples. It is validated for Immunohistochemistry
Specifications
Specifications
| Antigen | Cerebellin-4 |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CBLNL1, cerebellin 4 precursor, cerebellin precursor-like 1, cerebellin-4, Cerebellin-like glycoprotein 1, dJ885A10.1 |
| Host Species | Sheep |
| Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Cerebellin-4, Gln28-Leu201, Accession # Q9NTU7 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?