missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cerebellin 3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559498
This item is not returnable.
View return policy
Description
Immunogen sequence: GSEPVLLEGE CLVVCEPGRA AAGGPGGAAL GEAPPGRVAF AAVRSHHHEP AGETGNGTSG AIYFDQV Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.
Members of the precerebellin family, such as CBLN3, contain a cerebellin motif (see CBLN1; MIM 600432) and a C-terminal C1q signature domain (see MIM 120550) that mediates trimeric assembly of atypical collagen complexes. However, precerebellins do not contain a collagen motif, suggesting that they are not conventional components of the extracellular matrix (Pang et al., 2000 [PubMed 10964938]).[supplied by OMIM, Aug 2009].
Specifications
| Cerebellin 3 | |
| Polyclonal | |
| Unconjugated | |
| CBLN3 | |
| CBLN3; cerebellin 3 precursor; cerebellin 3 precursor protein; cerebellin-3; precerebellin 3; PRO1486; RGD1563731; UNQ755/PRO1486 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 643866 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UW01 | |
| CBLN3 | |
| Recombinant protein corresponding to Human Cerebellin 3. Recombinant protein control fragment (Product #RP-98450). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction