missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerebellin-1/Precerebellin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£141.00
Specifications
| Antigen | Cerebellin-1/Precerebellin |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2-4 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
Cerebellin-1/Precerebellin Polyclonal specifically detects Cerebellin-1/Precerebellin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry.Specifications
| Cerebellin-1/Precerebellin | |
| Western Blot 2-4 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| P23435 | |
| 869 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy- terminus human precerebellin. The immunogen is located within the last 50 amino acids of Precerebellin. Amino Acid Squence: QMEKGDRAYLKLERGN | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| cerebellin 1 precursor, cerebellin-1, CLN1, precerebellin | |
| CBLN1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title