missing translation for 'onlineSavingsMsg'
Learn More

Cerebellin-1/Precerebellin Antibody - BSA Free, Novus Biologicals™

Product Code. p-200070724 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
0.03mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18713034 0.025 mg 0.03mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18713034 Supplier Novus Biologicals Supplier No. NBP1772390.025mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 1 publication

Cerebellin-1/Precerebellin Polyclonal specifically detects Cerebellin-1/Precerebellin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cerebellin-1/Precerebellin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 2 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P23435
Gene Alias cerebellin 1 precursor, cerebellin-1, CLN1, precerebellin
Gene Symbols CBLN1
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from the middle of human precerebellin. The immunogen is located within amino acids 50 - 100 of Precerebellin. Amino Acid Squence: IRSTNHEPSEMSNRT
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 869
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.