missing translation for 'onlineSavingsMsg'
Learn More

Cercosporamide, MedChemExpress

Product Code. 16431659
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16431659 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16431659 Supplier Medchem Express Supplier No. HY16982
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cercosporamide is a highly potent, ATP-competitive Pkc1 kinase inhibitor, with an IC50 of Ki of Mnk inhibitor.

Cercosporamide is a highly potent, ATP-competitive Pkc1 kinase inhibitor, with an IC50 of Ki of Mnk inhibitor.

Specifications

Chemical Name or Material Cercosporamide
CAS 131436-22-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H13NO7
Quantity 1 mg
Synonym (-)-Cercosporamide
SMILES O=C(C1=C(C([C@@]23C)=C(O)C=C1O)OC2=CC(O)=C(C(C)=O)C3=O)N
Molecular Weight (g/mol) 331.28
Formula Weight 331.28
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.