missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Cercosporamide
Click to view available options
Quantity:
331 μg
Unit Size:
331µg
Description
Cercosporamide is a potent inhibitor of MAP-kinase interacting kinase-2 (Mnk2) and JAK3 (IC50 values are 11, 31 and 116 nM for Mnk2, JAK3 and Mnk1 respectively). Blocks eIF4E phosphorylation; exhibits antiproliferative and proapoptotic activity in cancer cells in vitro. Also inhibits Pkc1. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | (9aS)-8-Acetyl-9,9a-dihydro-1,3,7-trihydroxy-9a-methyl-9-oxo-4-dibenzofurancaboxamide |
| CAS | 131436-22-1 |
| Quantity | 331 μg |
| Target | Mnk Inhibitors |
| Molecular Formula | C16H13NO7 |
| Purity | 96% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction