missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CERCAM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565673
This item is not returnable.
View return policy
Description
Immunogen sequence: FDQHPNEQYKA HFWPRDLVAF SAQPLLAAPT HYAGDAEWLS DTETSSPWDD D Highest antigen sequence identity to the following orthologs - mouse 87%, rat 88%.
Probable cell adhesion protein involved in leukocyte transmigration across the blood-brain barrier. Does not express any beta-galactosyltransferase activity in vitro. [UniProt].
Specifications
| CERCAM | |
| Polyclonal | |
| Unconjugated | |
| CERCAM | |
| 2600006K01Rik; AL024097; CEECAM1; CERCAM; cerebral cell adhesion molecule; cerebral endothelial cell adhesion molecule; cerebral endothelial cell adhesion molecule 1; GLT25D3; glycosyltransferase 25 domain containing 3; glycosyltransferase 25 family member 3; Inactive glycosyltransferase 25 family member 3; KIAA1502; Probable inactive glycosyltransferase 25 family member 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51148 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5T4B2 | |
| CERCAM | |
| Recombinant Human CERCAM. Recombinant protein control fragment (Product #RP-106347). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction