missing translation for 'onlineSavingsMsg'
Learn More

Cerberus 1 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18322535 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322535 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322535 Supplier Novus Biologicals Supplier No. NBP299621UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cerberus 1 Polyclonal specifically detects Cerberus 1 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cerberus 1
Applications ELISA
Classification Polyclonal
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias cerberus, cerberus 1 (Xenopus laevis) homolog (cysteine knot superfamily), cerberus 1, cysteine knot superfamily, homolog (Xenopus laevis), cerberus-related 1, Cerberus-related protein, DAN domain family member 4, DAND4, MGC119894, MGC119895, MGC96951
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Cerberus 1 (AAH69371.1; Met1-Ala267).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 9350
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.