missing translation for 'onlineSavingsMsg'
Learn More

Ceranib 1, Tocris Bioscience™

Product Code. 18025962
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18025962

Brand: Tocris Bioscience 4448/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ceramidase inhibitor; antiproliferative

Ceramidase inhibitor; inhibits proliferation in SKOV3 ovarian carcinoma cells (IC50 = 3.9 μM). Induces accumulation of ceramide species and decreases sphingosine and sphingosine-1-phosphate (S1P) levels in SKOV3 cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328076-61-5
Molecular Formula C26H21NO3
Molecular Weight (g/mol) 395.458
InChI Key OJMCCNQNCSCAJV-RVDMUPIBSA-N
Synonym ceranib 1, 3-3-4-methoxy-phenyl-acryloyl-6-methyl-4-phenyl-1h-quinolin-2-one, 3-2e-3-4-methoxyphenyl-1-oxo-2-propen-1-yl-6-methyl-4-phenyl-2 1h-quinolinone, 3-2e-3-4-methoxyphenyl prop-2-enoyl-6-methyl-4-phenyl-1h-quinolin-2-one, 3-e-3-4-methoxyphenyl prop-2-enoyl-6-methyl-4-phenyl-1h-quinolin-2-one
PubChem CID 5761166
IUPAC Name 3-[(E)-3-(4-methoxyphenyl)prop-2-enoyl]-6-methyl-4-phenyl-1H-quinolin-2-one
SMILES CC1=CC2=C(C=C1)NC(=O)C(=C2C3=CC=CC=C3)C(=O)C=CC4=CC=C(C=C4)OC

Specifications

Quantity 10 mg
Formula Weight 395.45
Percent Purity >98%
Chemical Name or Material Ceranib 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.