missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ceramide
GREENER_CHOICE

Product Code. 30001121
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001121 50 mg 50mg
30001120 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001121 Supplier Tocris Bioscience™ Supplier No. 0744/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ser/Thr protein phosphatase activator

A potent modulator of cell proliferation and differentiation. Activates protein phosphatase-1 (PP1) and -2A (PP2A), as well as ceramide-activated protein phosphatase (CAPP) in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3102-57-6
Molecular Formula C20H39NO3

Specifications

Quantity 50 mg
Formula Weight 341.53
Purity >98%
Molecular Weight (g/mol) 341.54
Chemical Name or Material Ceramide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.