Learn More
Novus Biologicals™ Ceramide Kinase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30874-25ug
 View more versions of this product
Product Code. 30067070
  • £161.00 / 25µg
Description
Ceramide Kinase Polyclonal antibody specifically detects Ceramide Kinase in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ceramide Kinase
Polyclonal
Western Blot : 1-10 μg/mL, ELISA
ceramide kinase, dA59H18.2, DKFZp434E0211, EC 2.7.1.138, FLJ21430, FLJ23239, hCERKLipid kinase 4, KIAA1646dA59H18.3, lipid kinase LK4, LK4Acylsphingosine kinase, MGC131878
Synthetic peptide derived from human Ceramide Kinase
25 μg
Apoptosis, Cell Biology, Lipid and Metabolism
64781
Store at -20°C. Avoid freeze/thaw cycles.
ELISA, Western Blot
Unconjugated
Phosphate buffered saline
Rabbit
Ammonium sulfate precipitation
RUO
Primary
Human
Purified