missing translation for 'onlineSavingsMsg'
Learn More

Ceramide Kinase Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18640091 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18640091 0.02 mL 0.02mL
18677220 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18640091 Supplier Novus Biologicals Supplier No. NBP2922710.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Ceramide Kinase Polyclonal antibody specifically detects Ceramide Kinase in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ceramide Kinase
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias ceramide kinase, dA59H18.2, DKFZp434E0211, EC 2.7.1.138, FLJ21430, FLJ23239, hCERKLipid kinase 4, KIAA1646dA59H18.3, lipid kinase LK4, LK4Acylsphingosine kinase, MGC131878
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 400 to the C-terminus of human CERK (NP_073603.2). CRRSPRGLSPAAHLGDGSSDLILIRKCSRFNFLRFLIRHTNQQDQFDFTFVEVYRVKKFQFTSKHMEDEDSDLKEGGKKRFGHICSSHPSCCCTVSNSSWNCDGEVLHSPAIEVRVHCQLVRLFARGIEENPKPDSHS
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 64781
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.