missing translation for 'onlineSavingsMsg'
Learn More

Cephapirin Sodium, TRC

Product Code. 30135794
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135794 100 mg 100mg
17964545 25 mg 25mg
17930475 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135794 Supplier Toronto Research Chemicals Supplier No. TRCC261510100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24356-60-3
Molecular Formula C17 H16 N3 O6 S2 . Na
Molecular Weight (g/mol) 445.45
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-(hydroxymethyl)-8-oxo-7-[2-(4-pyridylthio)acetamido]-, acetate (ester), monosodium salt (8CI), 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[(acetyloxy)methyl]-8-oxo-7-[[(4-pyridinylthio)acetyl]amino]-, monosodium salt, (6R,7R)- (9CI), 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[(acetyloxy)methyl]-8-oxo-7-[[(4-pyridinylthio)acetyl]amino]-, monosodium salt, (6R-trans)-, 7-[α-(4-Pyridylthio)acetamido]cephalosporanic acid sodium salt, Ambrocef, BLP 1322, Brisfirina, Bristocef, Cef-Lak, Cefa-Lak, Cefadyl, Cefaloject, Cefapirin sodium, Cefaprin sodium, Cefatrexyl, Cephapirin sodium, Cephatrexil, NSC 179171, Piricef, Sodium cefapirin, Sodium cephapirin, Today
IUPAC Name sodium;(6R,7R)-3-(acetyloxymethyl)-8-oxo-7-[(2-pyridin-4-ylsulfanylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES [Na+].CC(=O)OCC1=C(N2[C@H](SC1)[C@H](NC(=O)CSc3ccncc3)C2=O)C(=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H17N3O6S2.Na/c1-9(21)26-6-10-7-28-16-13(15(23)20(16)14(10)17(24)25)19-12(22)8-27-11-2-4-18-5-3-11;/h2-5,13,16H,6-8H2,1H3,(H,19,22)(H,24,25);/q;+1/p-1/t13-,16-;/m1./s1
Formula Weight 445.0378
Percent Purity >95%
Chemical Name or Material Cefapirin sodium
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.