missing translation for 'onlineSavingsMsg'
Learn More

Cephalothin sodium salt, Thermo Scientific™

A first-generation cephalosporin antibiotic

£66.29 - £248.00

Chemical Identifiers

CAS 58-71-9
Molecular Formula C16H15N2NaO6S2
Molecular Weight (g/mol) 418.414
MDL Number MFCD00072025
InChI Key VUFGUVLLDPOSBC-XRZFDKQNSA-M
Synonym 7-(2-Thienylacetamido)cephalosporanic acid sodium salt; Cephalotin sodium salt
PubChem CID 23675321
ChEBI CHEBI:3542
IUPAC Name sodium;(6R,7R)-3-(acetyloxymethyl)-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES CC(=O)OCC1=C(N2C(C(C2=O)NC(=O)CC3=CC=CS3)SC1)C(=O)[O-].[Na+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15416999
SDS View Documents
Thermo Scientific Alfa Aesar
J65607.03
1g
ONLINE EXCLUSIVE PRICE
£67.80
£66.29 / 1g
Save £1.51
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15426999
SDS View Documents
Thermo Scientific Alfa Aesar
J65607.06
5g
ONLINE EXCLUSIVE PRICE
£253.00
£248.00 / 5g
Save £5.00
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Cephalothin sodium salt is used to study the mechanism of liposome encapsulated antibiotics1, strategies for co-opting β-lactamases of Gram-negative bacteria for treatment of antibiotics2, and for immunology studies in relation to antibiotics.3 It is used to study the effect of expression, binding and inhibition of penicillin-binding proteins especially PBP6 on bacterial cell wall mucopeptide synthesis.

Solubility
Soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

58-71-9
418.414
VUFGUVLLDPOSBC-XRZFDKQNSA-M
23675321
sodium;(6R,7R)-3-(acetyloxymethyl)-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
C16H15N2NaO6S2
MFCD00072025
7-(2-Thienylacetamido)cephalosporanic acid sodium salt; Cephalotin sodium salt
CHEBI:3542
CC(=O)OCC1=C(N2C(C(C2=O)NC(=O)CC3=CC=CS3)SC1)C(=O)[O-].[Na+]

Specifications

58-71-9
White
MFCD00072025
14,1982
Soluble in water
CC(=O)OCC1=C(N2C(C(C2=O)NC(=O)CC3=CC=CS3)SC1)C(=O)[O-].[Na+]
418.414
CHEBI:3542
Cephalothin sodium salt
Odorless
C16H15N2NaO6S2
4120706
7-(2-Thienylacetamido)cephalosporanic acid sodium salt; Cephalotin sodium salt
VUFGUVLLDPOSBC-XRZFDKQNSA-M
sodium;(6R,7R)-3-(acetyloxymethyl)-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
23675321
418.42
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c

H315-H317-H319-H334-H335

missing translation for 'einecsNumber' : 200-394-6

missing translation for 'rtecsNumber' : XI0388300

missing translation for 'tsca' : No

Recommended Storage : Keep cold

Product Identifier
  • Cephalothin sodium salt

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Cefalotin sodium

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.