missing translation for 'onlineSavingsMsg'
Learn More

Cephaloglycin, TRC

Product Code. 17984545
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17984545 500 mg 500mg
17933695 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17984545 Supplier Toronto Research Chemicals Supplier No. TRCC258000500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3577-01-3
Molecular Formula C18 H19 N3 O6 S
Molecular Weight (g/mol) 405.43
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[(acetyloxy)methyl]-7-[[(2R)-2-amino-2-phenylacetyl]amino]-8-oxo-, (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[(acetyloxy)methyl]-7-[(aminophenylacetyl)amino]-8-oxo-, [6R-[6α,7β(R*)]]-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[(acetyloxy)methyl]-7-[[(2R)-aminophenylacetyl]amino]-8-oxo-, (6R,7R)- (9CI), 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-(2-amino-2-phenylacetamido)-3-(hydroxymethyl)-8-oxo-, acetate (ester), D- (8CI), (6R,7R)-3-[(Acetyloxy)methyl]-7-[[(2R)-2-amino-2-phenylacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-(2-D-α-Aminophenylacetamido)cephalosporanic acid, Cefaloglycin, Cefaloglycine, Cephaloglycin, Cephaloglycine, D-(-)-Cephaloglycin, D-Cephaloglycine, Kafocin, Kefglycin, Lilly 39435;
IUPAC Name (6R,7R)-3-(acetyloxymethyl)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CC(=O)OCC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)c3ccccc3)C2=O)C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C18H19N3O6S/c1-9(22)27-7-11-8-28-17-13(16(24)21(17)14(11)18(25)26)20-15(23)12(19)10-5-3-2-4-6-10/h2-6,12-13,17H,7-8,19H2,1H3,(H,20,23)(H,25,26)/t12-,13-,17-/m1/s1
Formula Weight 405.0995
Chemical Name or Material Cefaloglycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.