missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Cephalexin, Monohydrate
Description
Cephalexin Monohydrate is a cephalosporin antibiotic. Cephalexin, as a cephalosporin, inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs).
This interference disrupts the cross-linking of peptidoglycans in the bacterial cell wall, leading to structural instability and eventual cell lysis. Cephalexin is sometimes used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion in culture medium allows for the specific growth of bacteria lacking resistance to cephalexin.
Specifications
Specifications
| Chemical Name or Material | Cephalexin |
| CAS | 23325-78-2 |
| Quantity | 5 g |
| Molecular Formula | C16H17N3O4S-H2O |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?