missing translation for 'onlineSavingsMsg'
Learn More

Cephalexin hydrate, 97+%

Product Code. 15455439
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455439 5 g 5g
15465439 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455439 Supplier Thermo Scientific Alfa Aesar Supplier No. J63172.06
  • ONLINE EXCLUSIVE PRICE
    £66.50
    £53.65 / 5g
    Save £12.85
    19% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cephalexin hydrate, cefalexin hydrate, CAS # 23325-78-2, is a first-generation cephalosporin antibiotic from the beta-lactam family. | CAS: 15686-71-2 | C16H20ClN3O5S | 401.862 g/mol

Cephalexin is a cephalosporin antibiotic used to study the effect of expression, binding, and inhibition of PBP3 and other penicillin-binding proteins (PBPs) on bacterial cell wall mucopeptide synthesis. Cephalexin disrupts the synthesis of the peptidoglycan layer of bacterial cell walls which is responsible for cell wall structural integrity. Peptidoglycan synthesis is facilitated by transpeptidases known as penicillin-binding proteins (PBPs). PBPs bind to the D-Ala-D-Ala at the end of muropeptides (peptidoglycan precursors) to crosslink the peptidoglycan. Cephalexin antibiotics mimic the D-Ala-D-Ala site, thereby competitively inhibiting PBP crosslinking of peptidoglycan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Cephalexin hydrate is the hydrate of the cephalosporin antibiotic, cephalexin

• Cephalexin is a beta-lactam antibiotic that inhibits bacterial growth and division

• Cephalexin binds to penicillin-binding protein and inhibits cell wall synthesis

Application

• Cephalexin works best on gram-positive microbes

• Useful for immunology research, particularly on antibiotic allergies and cross-reactivity

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15686-71-2
Molecular Formula C16H20ClN3O5S
Molecular Weight (g/mol) 401.862
MDL Number MFCD00167148
InChI Key YHJDZIQOCSDIQU-OEDJVVDHSA-N
PubChem CID 57515946
IUPAC Name (6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydron;chloride;hydrate
SMILES [H+].CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=CC=C3)N)SC1)C(=O)O.O.[Cl-]

Specifications

Type Cephalexin Hydrate
Physical Form Powder
Color White
Quantity 5 g
Beilstein 965503
Merck Index 141974
Solubility Information Soluble in DMSO and Methanol.
Formula Weight 347.39 (Anhydrous)
Percent Purity ≥97%
Chemical Name or Material Cephalexin hydrate
compliance-icons
Product Identifier
  • Cephalexin hydrate
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: Cephalexin hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.