missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CEP97 Polyclonal antibody specifically detects CEP97 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | CEP97 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 550 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 2810403B08Rik, centrosomal protein 97kDa, centrosomal protein of 97 kDa, Cep97, FLJ23047, FLJ26462, Leucine-rich repeat and IQ domain-containing protein 2, leucine-rich repeats and IQ motif containing 2, LRRIQ2 |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 625 and 675 of human centrosomal protein 97kDa using the numbering given in entry NP_078824.2 (GeneID 79598). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?