missing translation for 'onlineSavingsMsg'
Learn More

CEP97 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18425734 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425734 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425734 Supplier Novus Biologicals Supplier No. NBP106508UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CEP97 Polyclonal antibody specifically detects CEP97 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP97
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias 2810403B08Rik, centrosomal protein 97kDa, centrosomal protein of 97 kDa, Cep97, FLJ23047, FLJ26462, Leucine-rich repeat and IQ domain-containing protein 2, leucine-rich repeats and IQ motif containing 2, LRRIQ2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 625 and 675 of human centrosomal protein 97kDa using the numbering given in entry NP_078824.2 (GeneID 79598).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79598
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.