missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP95 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49042-25ul
This item is not returnable.
View return policy
Description
CEP95 Polyclonal antibody specifically detects CEP95 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CEP95 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| CCDC45, Centrosomal Protein 95kDa, CEP45, Cep95, Coiled-Coil Domain Containing 45, Coiled-Coil Domain-Containing Protein 45 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ALRRHDLLTTLVKKEYEHNKRLQDFKDCIRRQRLTQSKIKENRQQIVRARKYYDDYRVQLCAKMMRMRTREEMIFKKLFEEGLNIQKQRLRDLRNYAKEKRDEQRRRHQDELDSM | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 90799 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction