missing translation for 'onlineSavingsMsg'
Learn More

CEP76 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18423094 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423094 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423094 Supplier Novus Biologicals Supplier No. NBP128749JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CEP76 Polyclonal antibody specifically detects CEP76 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP76
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias C18orf9, centrosomal protein 76kDa, centrosomal protein of 76 kDa, Cep76, chromosome 18 open reading frame 9, FLJ12542, HsT1705
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1 and 30 of human centrosomal protein 76kDa using the numbering given in entry NP_079175.2 (GeneID 79959).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79959
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.