missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP76 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-28749R
This item is not returnable.
View return policy
Description
CEP76 Polyclonal antibody specifically detects CEP76 in Human samples. It is validated for Western Blot
Specifications
| CEP76 | |
| Polyclonal | |
| Western Blot | |
| C18orf9, centrosomal protein 76kDa, centrosomal protein of 76 kDa, Cep76, chromosome 18 open reading frame 9, FLJ12542, HsT1705 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 30 of human centrosomal protein 76kDa using the numbering given in entry NP_079175.2 (GeneID 79959). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 79959 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction