missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEP68 Monoclonal Antibody (OTI1F6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526304
This item is not returnable.
View return policy
Description
CEP68 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Centrosomal protein of 68 kDa is a protein that in humans is encoded by the CEP68 gene. It is mapped to chromosome 2. CEP68 is required for centrosome cohesion. It decorates fibres emanating from the proximal ends of centrioles. CEP68 and rootletin depend both on each other for centriole association, and both also require CEP250 for their function.
Specifications
| CEP68 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q76N32 | |
| CEP68 | |
| Full length human recombinant protein of CEP68 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1F6 | |
| Unconjugated | |
| CEP68 | |
| 6030463E10Rik; AI481761; BC027174; centrosomal protein 68; centrosomal protein 68kDa; centrosomal protein of 68 kDa; Cep68; Kiaa0582; RGD1309101 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23177 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction