missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEP63 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103728
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CEP63.
CEP63 is a protein with six coiled-coil domains. The protein is localized to the centrosome, a non-membranous organelle that functions as the major microtubule-organizing center in animal cells.
Specifications
| CEP63 | |
| Polyclonal | |
| Unconjugated | |
| CEP63 | |
| 4921501M07; AL450317.13gm1; AW107703; CD20R; centrosomal protein 63; centrosomal protein 63kDa; centrosomal protein of 63 kDa; centrosome protein Cep63; CEP63; D9Mgc41; D9Mgc48e; ET2; LOW QUALITY PROTEIN: centrosomal protein of 63 kDa; SCKL6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 28135, 80254 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3UPP8, Q96MT8 | |
| CEP63 | |
| A synthesized peptide derived from human CEP63(Accession Q96MT8), corresponding to amino acid residues D539-I589. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction