Learn More
CEP63 Mouse anti-Human, Unconjugated, Clone: 2G12B2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66996-1-IG-20UL
This item is not returnable.
View return policy
Description
The CEP63 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CEP63. This antibody recognizes human, mouse, rat antigen. The CEP63 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CEP63 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, ELISA | |
| 2G12B2 | |
| Unconjugated | |
| 20 μL | |
| 80254 | |
| IgG1 |