missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEP63 Monoclonal Antibody (OTI1C7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527116
This item is not returnable.
View return policy
Description
CEP63 Monoclonal Antibody for Western Blot, IHC (P)
CEP63 is a protein with six coiled-coil domains. The protein is localized to the centrosome, a non-membranous organelle that functions as the major microtubule-organizing center in animal cells.
Specifications
| CEP63 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96MT8 | |
| CEP63 | |
| Full length human recombinant protein of CEP63 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C7 | |
| Unconjugated | |
| CEP63 | |
| 4921501M07; AL450317.13gm1; AW107703; CD20R; centrosomal protein 63; centrosomal protein 63kDa; centrosomal protein of 63 kDa; centrosome protein Cep63; CEP63; D9Mgc41; D9Mgc48e; ET2; LOW QUALITY PROTEIN: centrosomal protein of 63 kDa; SCKL6 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 80254 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction