missing translation for 'onlineSavingsMsg'
Learn More

CEP63 Antibody (OTI2H2), DyLight 594, Novus Biologicals™

Product Code. 18405407 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405407 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405407 Supplier Novus Biologicals Supplier No. NBP272221DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CEP63 Monoclonal antibody specifically detects CEP63 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP63
Applications Western Blot
Classification Monoclonal
Clone OTI2H2
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias centrosomal protein 63kDa, centrosomal protein of 63 kDa, centrosome protein CEP63, Cep63, FLJ13386, MGC78416
Host Species Mouse
Immunogen Full length recombinant protein of human CEP63 (NP_001035859) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Direct Reversal of DNA Damage
Primary or Secondary Primary
Gene ID (Entrez) 80254
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.