Learn More
Invitrogen™ CEP57L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114061
Description
Immunogen sequence: DIECELECLL KKMEIKGEQI SKLKKHQDSV CKLQQKVQNS KMSEASGIQQ For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 79%; Dog: 85%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.
C6orf182 also named as Centrosomal protein of 57 kDa related protein is a 460 amino-acid protein, which belongs to translokin family. C6orf182 as centrosomal protein may be required for microtubule attachment to centrosomes and localizes in the cytoplasm. The function of C6orf182 need to be further studied.
Specifications
| CEP57L1 | |
| Polyclonal | |
| Unconjugated | |
| CEP57L1 | |
| 2410017P07Rik; 4930484D11Rik; AI607327; bA487F23.2; C6orf182; centrosomal protein 57 like 1; centrosomal protein 57kDa-like 1; centrosomal protein 57kDa-like protein 1; centrosomal protein 57-like 1; Centrosomal protein CEP57L1; centrosomal protein of 57 kDa-related protein; CEP57L1; Cep57R; Cep57-related protein; RGD1311358 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 285753 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IYX8 | |
| CEP57L1 | |
| Synthetic peptide directed towards the middle region of human C6orf182. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.