missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP57L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | CEP57L1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
CEP57L1 Polyclonal specifically detects CEP57L1 in Human samples. It is validated for Western Blot.Specifications
| CEP57L1 | |
| Polyclonal | |
| Rabbit | |
| centrosomal protein 57kDa-like 1, centrosomal protein 57kDa-like protein 1 | |
| CEP57L1 | |
| IgG | |
| 54 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 285753 | |
| Synthetic peptides corresponding to C6ORF182 The peptide sequence was selected from the middle region of C6ORF182. Peptide sequence DIECELECLLKKMEIKGEQISKLKKHQDSVCKLQQKVQNSKMSEASGIQQ. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title