missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP57L1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92923-0.02ml
This item is not returnable.
View return policy
Description
CEP57L1 Polyclonal antibody specifically detects CEP57L1 in Human samples. It is validated for Western Blot
Specifications
| CEP57L1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| centrosomal protein 57kDa-like 1, centrosomal protein 57kDa-like protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-167 of human CEP57L1 (NP_776191.1). MDSELMHSIVGSYHKPPERVFVPSFTQNEPSQNCHPANLEVTSPKILHSPNSQALILALKTLQEKIHRLELERTQAEDNLNILSREAAQYKKALENETNERNLAHQELIKQKKDISIQLSSAQSRCTLLEKQLEYTKRMVLNVEREKNMILEQQAQLQREKEQDQMK | |
| 0.02 mL | |
| Cell Biology, Cell Cycle and Replication | |
| 285753 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction