missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP57 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15863
This item is not returnable.
View return policy
Description
CEP57 Polyclonal specifically detects CEP57 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CEP57 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| centrosomal protein 57kDa, Cep57, FGF2-interacting protein, KIAA0092centrosomal protein of 57 kDa, PIG8, Testis-specific protein 57, Translokin, TSP57proliferation-inducing protein 8 | |
| Rabbit | |
| 57 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Expected to cross react based on sequence identity: mouse (94%), Rat (92%), cow (95%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 0.94mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q86XR8 | |
| CEP57 | |
| Recombinant protein encompassing a sequence within the center region of human CEP57. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 9702 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction