missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP55 Rabbit anti-Human, Mouse, Rat, Clone: S05-6A6, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-15012-100UL
This item is not returnable.
View return policy
Description
CEP55 Monoclonal antibody specifically detects CEP55 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CEP55 | |
| Monoclonal | |
| Unconjugated | |
| C10orf3, cancer/testis antigen 111, centrosomal protein 55kDa, centrosomal protein of 55 kDa, Cep55, chromosome 10 open reading frame 3, CT111, FLJ10540, Up-regulated in colon cancer 6, URCC6 | |
| A synthetic peptide of human CEP55 (Uniprot#: Q53EZ4) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S05-6A6 | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| Rabbit | |
| Affinity-purified | |
| RUO | |
| 55165 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction