missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP55 Antibody (OTI10C4), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71990AF750
This item is not returnable.
View return policy
Description
CEP55 Monoclonal specifically detects CEP55 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CEP55 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CEP55 | |
| Recombinant protein fragment corresponding to amino acids 1-280 of human CEP55 (NP_060601) produced in E.coli. | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI10C4 | |
| Western Blot, Immunohistochemistry | |
| C10orf3, cancer/testis antigen 111, centrosomal protein 55kDa, centrosomal protein of 55 kDa, Cep55, chromosome 10 open reading frame 3, CT111, FLJ10540, Up-regulated in colon cancer 6, URCC6 | |
| Mouse | |
| 0.1 mL | |
| 55165 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction