missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CEP44 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16581772
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16581772 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16581772 Supplier Invitrogen™ Supplier No. PA598953

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CEP44 Polyclonal Antibody for Western Blot, IHC (P), ELISA

CEP44 (Centrosomal Protein 44) is a Protein Coding gene. Diseases associated with CEP44 include Retinitis Pigmentosa 29. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEP44
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CEP44
Gene Accession No. Q9C0F1
Gene Alias 4933440G23Rik; centrosomal protein 44; centrosomal protein 44kDa; centrosomal protein of 44 kDa; Cep44; HBV PreS1-transactivated protein 3; KIAA1712; mKIAA1712; PS1TP3
Gene Symbols CEP44
Host Species Rabbit
Immunogen Recombinant Human Centrosomal protein of 44 kDa protein (1-280AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80817
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.