missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP44 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92537-0.1ml
This item is not returnable.
View return policy
Description
CEP44 Polyclonal antibody specifically detects CEP44 in Human, Mouse samples. It is validated for Western Blot
Specifications
| CEP44 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| centrosomal protein 44kDa, centrosomal protein of 44 kDa, KIAA1712, PS1TP3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human CEP44 (NP_001035247.1). MATGDLKRSLRNLEQVLRLLNYPEEVDCVGLIKGDPAASLPIISYSFTSYSPYVTELIMESNVELIAKNDLRFIDAVYKLLRDQFNYKPILTKKQFIQCGFAEWKIQIVCDILNCVMKKHKELSSLQKIPSQQRKKISSGKSEPPLGNEKISAEAVGVDISGRFMTSGKKKAVVIRHLYNEDNVDISEDTLSPITDVNEAVDVSDLNATEIKMPEVKVPEIKAEQQDVNVNPEITALQTMLAECQENLKKLTSIEKRLDCLEQKMKGKVMVDENTWTNLL | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication | |
| 80817 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction