missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEP350 Monoclonal Antibody (CL3423)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531385
This item is not returnable.
View return policy
Description
Immunogen sequence: QVVQSQREVT EVLQEATCKI AAQQSETARL TTDAARQICE MAELTRTHIS DAVVASGAPL AILYDHQRQH LPDFVKQLRT RTETDRKSPS VSLSQ Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 83%.
May be involved in transcriptional regulation.
Specifications
| CEP350 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5VT06 | |
| CEP350 | |
| Recombinant protein fragment of Human CEP350 (Product #RP-105842) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| CL3423 | |
| Unconjugated | |
| CEP350 | |
| 4933409L06Rik; 6430546F08Rik; Cap350; centrosomal protein 350; centrosomal protein 350kDa; centrosome-associated protein 350; Centrosome-associated protein of 350 kDa; CEP350; GM133; Kiaa0480 | |
| Mouse | |
| Protein A | |
| RUO | |
| 9857 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction