missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEP290 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560550
This item is not returnable.
View return policy
Description
Immunogen sequence: QVESINKELE ITKEKLHTIE QAWEQETKLG NESSMDKAKK SITNSDIVSI SKKITMLEMK ELNERQRAEH CQKMYEHLRT SLKQMEERNF ELETKFAELT Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
This gene encodes a protein with 13 putative coiled-coil domains, a region with homology to SMC chromosome segregation ATPases, six KID motifs, three tropomyosin homology domains and an ATP/GTP binding site motif A. The protein is localized to the centrosome and cilia and has sites for N-glycosylation, tyrosine sulfation, phosphorylation, N-myristoylation, and amidation. Mutations in this gene have been associated with Joubert syndrome and nephronophthisis and the presence of antibodies against this protein is associated with several forms of cancer.
Specifications
| CEP290 | |
| Polyclonal | |
| Unconjugated | |
| Cep290 | |
| 3H11Ag; b2b1454Clo; b2b1752Clo; Bardet-Biedl syndrome 14 protein; Bardet-Biedl syndrome 14 protein homolog; BBS14; BC004690; Cancer/testis antigen 87; centrosomal protein 290; centrosomal protein 290kDa; centrosomal protein of 290 kDa; Cep290; CT87; CTCL tumor antigen se2-2; JBTS5; Kiaa0373; LCA10; Meckel syndrome, type 4; MKS4; monoclonal antibody 3H11 antigen; nephrocystin-6; nephrocytsin-6; Nphp6; POC3; POC3 centriolar protein homolog; prostate cancer antigen T21; rd16; RGD1311640; SLSN6; tumor antigen se2-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80184 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15078 | |
| Cep290 | |
| Recombinant protein corresponding to Human CEP290. Recombinant protein control fragment (Product #RP-97890). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction