missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEP250 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568141
This item is not returnable.
View return policy
Description
Predicted to react with Human samples.
Probably plays an important role in centrosome cohesion during interphase.
Specifications
| CEP250 | |
| Polyclonal | |
| Unconjugated | |
| CEP250 | |
| 250 kDa centrosomal protein; AW490617; B230210E21Rik; centrosomal Nek2-associated protein 1; Centrosomal protein 2; centrosomal protein 250; centrosomal protein 250kDa; centrosome associated protein; centrosome-associated protein CEP250; Cep2; CEP250; CNAP1; C-Nap1; Inmp; Intranuclear matrix protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11190 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BV73 | |
| CEP250 | |
| Synthesized peptide derived from human CEP250. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction