missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEP19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567148
This item is not returnable.
View return policy
Description
Immunogen sequence: QSLAETMEQIQ RETTIDPEED LNKLDDKELA KRKSIMDELF EKNQKKKDDP NFVYDIEVEF PQDDQLQSCG WDTESADEF Highest antigen sequence identity to the following orthologs - mouse 88%, rat 90%.
The protein encoded by this gene localizes to centrosomes and primary cilia and co-localizes with a marker for the mother centriole. This gene resides in a region of human chromosome 3 that is linked to morbid obesity. A homozygous knockout of the orthologous gene in mouse resulted in mice with morbid obesity, hyperphagy, glucose intolerance, and insulin resistance. Mutations in this gene cause morbid obesity and spermatogenic failure (MOSPGF). This gene has a pseudogene on human chromosome 2.
Specifications
| CEP19 | |
| Polyclonal | |
| Unconjugated | |
| CEP19 | |
| 1500031L02Rik; AI428934; AL022620; C3orf34; centrosomal protein 19; centrosomal protein 19kDa; centrosomal protein of 19 kDa; Cep19; HSD5; MOSPGF; RGD621352; testis specific protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84984 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96LK0 | |
| CEP19 | |
| Recombinant Human CEP19. Recombinant protein control fragment (Product #RP-107795). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction