Learn More
Invitrogen™ CEP170B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551473
Description
Immunogen sequence: EPTPASFFIG DQNGDAVLSR KPLAAPGDGE GLGQTAQPSP PARDGVYVSA NGRMVIQLRP GRSPEPDGPA PAFLRQESFT KEPASGPPAP GKPPHISSHP LLQDLAATRA ARMDFHSQDT HLILKETETA LAALEARLLS NSVDAECE Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 73%.
CEP170B plays a role in microtubule organization.
Specifications
| CEP170B | |
| Polyclonal | |
| Unconjugated | |
| CEP170B | |
| AI466840; AW555464; Centrosomal protein 170B; centrosomal protein of 170 kDa protein B; CEP170B; CEP170R; Cep170-related; FAM68C; KIAA0284; mKIAA0284; si:ch73-160l8.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 283638 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y4F5 | |
| CEP170B | |
| Recombinant protein corresponding to Human CEP170B. Recombinant protein control fragment (Product #RP-88986). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.