missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: QPLRAQKEMS PSPPAAQDPG GTALVSAREQ SSERQHHPLG PTDMGRGEPV RRSAIRRGHR PRGSLDWPSE ERGPVLAHLP SSDVMASNHE T Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 58%.
CEP170B plays a role in microtubule organization.
Specifications
Specifications
| Antigen | CEP170B |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CEP170B |
| Gene Accession No. | Q9Y4F5 |
| Gene Alias | AI466840; AW555464; Centrosomal protein 170B; centrosomal protein of 170 kDa protein B; CEP170B; CEP170R; Cep170-related; FAM68C; KIAA0284; mKIAA0284; si:ch73-160l8.1 |
| Gene Symbols | CEP170B |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
